4-tert-Butyl-o-xylene ≥98.0%
Leverantör: TCI
Synonyms: 1,2-Dimethyl-4-tert-butylbenzene, 1-tert-Butyl-3,4-dimethylbenzene
B1338-25G B1338-500G
TCIAB1338-25GEA 415 SEK
TCIAB1338-25G TCIAB1338-500G
4-tert-Butyl-o-xylene ≥98.0%
4-tert-Butyl-o-xylene
Formel: C₁₂H₁₈
MW: 162.27 g/mol
Kokpunkt: 211°C
Flampunkt: 82°C
MDL Number: MFCD00038277
CAS nummer: 7397-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96f18941f625b6ab362d240796886d93050e0756&vid=96f18941f625b6ab362d240796886d93050e0756&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %