3-Methoxypyrocatechol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2,3-Dihydroxyanisole, 3-Methoxycatechol, 3-Methoxybenzene-1,2-diol

M0524-25G M0524-500G
TCIAM0524-25GEA 535 SEK
TCIAM0524-25G TCIAM0524-500G
3-Methoxypyrocatechol ≥98.0%
3-Methoxypyrocatechol
Formel: C₇H₈O₃
MW: 140,14 g/mol
Kokpunkt: 164°C
Smältpunkt: 39…40 °C
Densitet: 1,2 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00002191
CAS nummer: 934-00-9
EINECS: 213-276-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=205ab466c30c83dfe7f38b979b3916ba77ce0f5f&vid=205ab466c30c83dfe7f38b979b3916ba77ce0f5f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Melting point 38.0 to 44.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR