1,2,3,4-Tetraphenylcyclopentadiene ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1,2,3,4-Tetraphenyl-1,3-cyclopentadiene

T1333-1G T1333-5G
TCIAT1333-1GEA 1400 SEK
TCIAT1333-1G TCIAT1333-5G
1,2,3,4-Tetraphenylcyclopentadiene ≥98.0%
1,2,3,4-Tetraphenylcyclopentadiene
Formel: C₂₉H₂₂
MW: 370,49 g/mol
Smältpunkt: 182…184 °C
Densitet: 1,24 g/cm³ (20 °C)
MDL Number: MFCD00001355
CAS nummer: 15570-45-3
EINECS: 239-619-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20a165f358329c58e24b5a84632418e1278aaed1&vid=20a165f358329c58e24b5a84632418e1278aaed1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 177.0 to 182.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR