1-Isopropylpiperazine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

I0745-25G I0745-5G
TCIAI0745-25GEA 1650 SEK
TCIAI0745-25G TCIAI0745-5G
1-Isopropylpiperazine ≥98.0% (by GC, titration analysis)
1-Isopropylpiperazine
Formel: C₇H₁₆N₂
MW: 128,22 g/mol
Kokpunkt: 178°C
Flampunkt: 54°C
MDL Number: MFCD00167971
CAS nummer: 4318-42-7
EINECS: 224-345-3
UN: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f9fa770df495b2345ef9cc046c86c8d94462a2d&vid=2f9fa770df495b2345ef9cc046c86c8d94462a2d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR