3,5-Dimethoxybenzylamine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: 3,5-Dimethoxybenzyl amine
D4025-25G D4025-5G
TCIAD4025-25GEA 3500 SEK
TCIAD4025-25G TCIAD4025-5G
3,5-Dimethoxybenzylamine ≥98.0% (by GC, titration analysis)
3,5-Dimethoxybenzylamine
Formel: C₉H₁₃NO₂
MW: 167,21 g/mol
Kokpunkt: 96°C
Storage Temperature: Kylskåp
MDL Number: MFCD00052813
CAS nummer: 34967-24-3
UN: 3259
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54511551efd75c11eea2910d7b46fc1f03a4bbe9&vid=54511551efd75c11eea2910d7b46fc1f03a4bbe9&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %