1-Adamantaneethanol ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 1-(2-Hydroxyethyl)adamantane, 1-(2-Hydroxyethyl)tricyclo[3.3.1.1(3,7)]decane, 2-(1-Adamantyl)ethanol
A1619-25G A1619-5G
TCIAA1619-25GEA 5275 SEK
TCIAA1619-25G TCIAA1619-5G
1-Adamantaneethanol ≥98.0%
1-Adamantaneethanol
Formel: C₁₂H₂₀O
MW: 180,29 g/mol
Kokpunkt: 270 °C (1013 hPa)
Smältpunkt: 98,5…100 °C
MDL Number: MFCD00074756
CAS nummer: 6240-11-5
EINECS: 228-353-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d3eea10a273dfc04c25f8027e929e893e2737af&vid=9d3eea10a273dfc04c25f8027e929e893e2737af&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 74.0 to 78.0 deg-C