1,3-Dimethylurea ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: N,N'-Dimethylurea
D0289-25G D0289-500G
TCIAD0289-25GEA 215 SEK
TCIAD0289-25G TCIAD0289-500G
1,3-Dimethylurea ≥98.0%
1,3-Dimethylurea
Formel: C₃H₈N₂O
MW: 88,11 g/mol
Kokpunkt: 270°C
Smältpunkt: 96 °C
Densitet: 1,142 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008286
CAS nummer: 96-31-1
EINECS: 202-498-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eecb01e113d8297ec5f4793ffa6963d2ccee1c53&vid=eecb01e113d8297ec5f4793ffa6963d2ccee1c53&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 102.0 to 108.0 deg-C
Solubility in Water almost transparency