N-Methylethylamine ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: N-Ethylmethylamine

E0729-25G E0729-5G
TCIAE0729-25GEA 3325 SEK
TCIAE0729-25G TCIAE0729-5G
N-Methylethylamine ≥98.0%
N-Methylethylamine
Formel: C₃H₉N
MW: 59,11 g/mol
Kokpunkt: 35°C
Densitet: 0,6893 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00009030
CAS nummer: 624-78-2
EINECS: 210-862-1
UN: 2733
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=086c6c7fd38a7c830b989fb155458f9e3b782a37&vid=086c6c7fd38a7c830b989fb155458f9e3b782a37&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR