Benzyl isocyanate ≥99.0%

Leverantör: TCI EUROPE
Fara

I0426-25G I0426-5G
TCIAI0426-25GEA 2200 SEK
TCIAI0426-25G TCIAI0426-5G
Benzyl isocyanate ≥99.0%
Benzyl isocyanate
Formel: C₈H₇NO
MW: 133,15 g/mol
Kokpunkt: 118°C
Densitet: 1,0838 g/cm³ (20 °C)
Flampunkt: 45°C
Storage Temperature: Kylskåp
MDL Number: MFCD00009701
CAS nummer: 3173-56-6
EINECS: 221-640-9
UN: 2478
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72196b89d9d720b91356238e6c3adac3a1dda33d&vid=72196b89d9d720b91356238e6c3adac3a1dda33d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR