Tetrametylurea ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
1,1,3,3-Tetrametylurea
T0158-25G
T0158-500G
TCIAT0158-25GEA
230
SEK
TCIAT0158-25G
TCIAT0158-500G
Tetrametylurea ≥98.0%
Tetrametylurea
Formel:
C₅H₁₂N₂O MW: 116.16 g/mol Kokpunkt: 177°C Smältpunkt: -1 Flampunkt: 75°C Storage Temperature: Omgivande |
MDL Number:
MFCD00008319 CAS nummer: 632-22-4 EINECS: 211-173-9 Merck Index: 14,09230 |
|
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Specific gravity (20/20) | 0.9670 to 0.9720 |
Appearance | Colorless to Light yellow to Light orange clear liquid |
Refractive index n20/D | 1.4490 to 1.4530 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...