Etyldimetylamin ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: DMEA
D1538-25ML D1538-500ML
TCIAD1538-25MLEA 215 SEK
TCIAD1538-25ML TCIAD1538-500ML
Etyldimetylamin ≥98.0% (by GC, titration analysis)
Etyldimetylamin
Formel: C₄H₁₁N
MW: 73.14 g/mol
Kokpunkt: 37°C
Smältpunkt: -140
Flampunkt: –25°C
MDL Number: MFCD00009039
CAS nummer: 598-56-1
EINECS: 209-940-8
UN: 2733
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f86f0977fc835435d77a0e5d50702dd07c91929b&vid=f86f0977fc835435d77a0e5d50702dd07c91929b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %