5-Hydroxyindol-3-ylacetic syra ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 5-Hydroxyindole-3-acetic acid, 2-(5-Hydroxy-1H-indol-3-yl)acetic acid

H0355-1G H0355-100MG
TCIAH0355-1GEA 3925 SEK
TCIAH0355-1G TCIAH0355-100MG
5-Hydroxyindol-3-ylacetic syra ≥98.0% (by HPLC, titration analysis)
5-Hydroxyindol-3-ylacetic syra
Formel: C₁₀H₉NO₃
MW: 191,19 g/mol
Smältpunkt: 166 °C
Storage Temperature: Frys
MDL Number: MFCD00005639
CAS nummer: 54-16-0
EINECS: 200-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6f7fa34bcde10f06db67b0b198e6ce4d7193efe&vid=e6f7fa34bcde10f06db67b0b198e6ce4d7193efe&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Amber to Dark purple powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR