1-Allyl-2-thiourea ≥98.0%
Leverantör: TCI
Fara
Synonyms: N-Allylthiourea, Tiosinamine, 1-Allyl-2-thiourea, Allyl thiourea, Rhodalline, Thiosinamine, N-Allylthiourea
A0220-25G A0220-500G A0220-100G
TCIAA0220-25GEA 365 SEK
TCIAA0220-25G TCIAA0220-500G TCIAA0220-100G
1-Allyl-2-thiourea ≥98.0%
1-Allyl-2-thiourea
Formel: H₂CCHCH₂NHC(S)NH₂
MW: 116.19 g/mol
Kokpunkt: 191
Smältpunkt: 74
Storage Temperature: Omgivande
MDL Number: MFCD00004940
CAS nummer: 109-57-9
EINECS: 203-683-5
UN: 2811
ADR: 6.1 III
Merck Index: 14,09362
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26bb5428d7ceee01da634ff63d3c6d7e182f03c1&vid=26bb5428d7ceee01da634ff63d3c6d7e182f03c1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (Volhard Method) min. 98.0 %
Melting point 69.0 to 73.0 deg-C
Solubility in Methanol almost transparency