Allyltrimethoxysilane 97,0 GC_ANALYSMETOD
Leverantör: TCI
|
Varning
|
Synonyms:
Trimethoxyallylsilane
A1504-25ML
A1504-5ML
TCIAA1504-25MLEA
1450
SEK
TCIAA1504-25ML
TCIAA1504-5ML
Allyltrimethoxysilane 97,0 GC_ANALYSMETOD
Allyltrimethoxysilane
Formel:
H₂CCHCH₂Si(OCH₃)₃ MW: 162.26 g/mol Kokpunkt: 136°C Flampunkt: 31°C |
MDL Number:
MFCD00053865 CAS nummer: 2551-83-9 EINECS: 219-855-8 UN: 1993 ADR: 3 III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid to slightly cloudy liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...