4-Chloro-α-methylbenzyl alcohol ≥97.0%
Leverantör: TCI
Synonyms:
1-(4-Chlorophenyl)ethanol, 4-Chlorophenyl methyl carbinol, p-Chloro-α-methylbenzyl alcohol
C0198-10G
C0198-25G
TCIAC0198-10GEA
615
SEK
TCIAC0198-10G
TCIAC0198-25G
4-Chloro-α-methylbenzyl alcohol ≥97.0%
4-Chloro-α-methylbenzyl alcohol
Formel:
C₈H₉ClO Kokpunkt: 136°C Densitet: 1,17 g/cm³ (20 °C) |
MDL Number:
MFCD00041038 CAS nummer: 3391-10-4 EINECS: 222-223-4 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...