(±)-4-Hydroxy-3-methoxymandelsyra ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Hydroxy-3-methoxy-DL-mandelic acid, VMA, Vanillyl mandelic acid, DL-4-Hydroxy-3-methoxymandelic acid, (±)-4-Hydroxy-3-methoxyphenylglycolic acid, 2-Hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid, Vanillylmandelic acid

H0268-1G H0268-5G
TCIAH0268-1GEA 535 SEK
TCIAH0268-1G TCIAH0268-5G
(±)-4-Hydroxy-3-methoxymandelsyra ≥98.0% (by HPLC, titration analysis)
(±)-4-Hydroxy-3-methoxymandelsyra
Formel: C₉H₁₀O₅
MW: 198,18 g/mol
Kokpunkt: 421 °C (1013 hPa)
Smältpunkt: 132…134 °C
Densitet: 1,44 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00004235
CAS nummer: 55-10-7
EINECS: 200-224-0
Merck Index: 14,09933
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0527df320b31b302ce2f757ac839f7c7840f87aa&vid=0527df320b31b302ce2f757ac839f7c7840f87aa&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR