(±)-Methyl mandelate ≥98.0%
Leverantör: TCI EUROPE
Synonyms: DL-Methyl glycolate, DL-2-Hydroxy-2-phenylacetic acid methyl ester, (±)-2-Hydroxy-2-phenylacetic acid methyl ester, (±)-Mandelic acid methyl ester, Methyl DL-mandelate, Methyl 2-hydroxy-2-phenylacetate, (±)-Methyl glycolate, DL-Mandelic acid methyl ester, DL-Methyl mandelate
M0040-25G M0040-500G
TCIAM0040-25GEA 295 SEK
TCIAM0040-25G TCIAM0040-500G
(±)-Methyl mandelate ≥98.0%
(±)-Methyl mandelate
Formel: C₉H₁₀O₃
MW: 166,18 g/mol
Kokpunkt: 250°C
Smältpunkt: 54…56 °C
Densitet: 1,1756 g/cm³ (20 °C)
MDL Number: MFCD00004493
CAS nummer: 4358-87-6
EINECS: 224-434-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cda819cf7a75deeb01f3d27c8cffa21a384826e1&vid=cda819cf7a75deeb01f3d27c8cffa21a384826e1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 54.0 to 57.0 deg-C
Solubility in Methanol almost transparency