Tetrahydrotiofen ≥99.0%

Leverantör: TCI EUROPE
Fara

Synonyms: THT

T0114-25ML T0114-500ML
TCIAT0114-25MLEA 230 SEK
TCIAT0114-25ML TCIAT0114-500ML
Tetrahydrotiofen ≥99.0%
Tetrahydrotiofen
Formel: C₄H₈S
MW: 88,17 g/mol
Kokpunkt: 121°C
Smältpunkt: –96 °C
Densitet: 1 g/cm³ (20 °C)
Flampunkt: 13°C
MDL Number: MFCD00005476
CAS nummer: 110-01-0
EINECS: 203-728-9
UN: 2412
ADR: 3,II
Merck Index: 12,09357
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd821b5bed9045d070f4a036bd9952feac0422e8&vid=cd821b5bed9045d070f4a036bd9952feac0422e8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR