2-Bromo-4-nitropyridine-N-oxide ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 2-Bromo-4-nitropyridine N-oxide, 2-Bromo-4-nitropyridine 1-oxide

B3145-1G B3145-5G
TCIAB3145-1GEA 495 SEK
TCIAB3145-1G TCIAB3145-5G
2-Bromo-4-nitropyridine-N-oxide ≥98.0%
2-Bromo-4-nitropyridine-N-oxide
Formel: C₅H₃BrN₂O₃
MW: 218,99 g/mol
MDL Number: MFCD00160743
CAS nummer: 52092-43-0
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21248a53eefc6ce6ebc352e969649917e2ce0bb7&vid=21248a53eefc6ce6ebc352e969649917e2ce0bb7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 144.0 to 148.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR