2-Methoxy-3-methylpyrazine ≥98.0%

Leverantör: TCI
Varning

M0810-25G M0810-5G
TCIAM0810-25GEA 670 SEK
TCIAM0810-25G TCIAM0810-5G
2-Methoxy-3-methylpyrazine ≥98.0%
2-Methoxy-3-methylpyrazine
Formel: C₆H₈N₂O
Kokpunkt: 87°C
Densitet: 1,07 g/cm³ (20 °C)
Flampunkt: 56°C
MDL Number: MFCD00006127
CAS nummer: 2847-30-5
EINECS: 220-651-6
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d284ce729f606d970245596a21a9f2ff2a074d22&vid=d284ce729f606d970245596a21a9f2ff2a074d22&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR