2-Iodopyrazine ≥97.0%

Leverantör: TCI
Varning

I0837-1G I0837-5G
TCIAI0837-1GEA 445 SEK
TCIAI0837-1G TCIAI0837-5G
2-Iodopyrazine ≥97.0%
2-Iodopyrazine
Formel: C₄H₃IN₂
Kokpunkt: 110°C
MDL Number: MFCD01319019
CAS nummer: 32111-21-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82a5659183f6b0d401b2b4094c243a68f0871f70&vid=82a5659183f6b0d401b2b4094c243a68f0871f70&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Orange to Brown clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR