1,4-Butanesultone ≥99.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1,4-Butane sultone
B0136-25G B0136-250G
TCIAB0136-25GEA 365 SEK
TCIAB0136-25G TCIAB0136-250G
1,4-Butanesultone ≥99.0%
1,4-Butanesultone
Formel: C₄H₈O₃S
MW: 136,17 g/mol
Kokpunkt: 146°C
Smältpunkt: 12,5…15 °C
Densitet: 1,331 g/cm³ (20 °C)
MDL Number: MFCD00006584
CAS nummer: 1633-83-6
EINECS: 216-647-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96b53df4aee4af05fcbe78e8e8e6e54eee8aa5cd&vid=96b53df4aee4af05fcbe78e8e8e6e54eee8aa5cd&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 99.0 %