2-Acetylnaphthalene ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2´-Acetonaphthone, 1-(Naphthalen-2-yl)ethanone, Methyl β-naphthyl ketone, 2-Acetonaphthone, 2'-Acetonaphthone
A0053-25G A0053-500G A0053-100G
TCIAA0053-25GEA 270 SEK
TCIAA0053-25G TCIAA0053-500G TCIAA0053-100G
2-Acetylnaphthalene ≥98.0%
2-Acetylnaphthalene
Formel: C₁₂H₁₀O
MW: 170,21 g/mol
Kokpunkt: 301°C
Smältpunkt: 52…54 °C
Densitet: 1,1171 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004108
CAS nummer: 93-08-3
EINECS: 202-216-2
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46c74c291cf0ed116133b04459c41366880ece6c&vid=46c74c291cf0ed116133b04459c41366880ece6c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 57.0 deg-C
Solubility in Methanol almost transparency