Dimetyl(1-naftyl)amin 99,0 GC, T
Leverantör: TCI
Varning
Synonyms: Dimethyl(1-naphthyl)amine, N,N-Dimethylnaphthalen-1-amine
D0755-25G D0755-500G
TCIAD0755-25GEA 1200 SEK
TCIAD0755-25G TCIAD0755-500G
Dimetyl(1-naftyl)amin 99,0 GC, T
Dimetyl(1-naftyl)amin
Formel: C₁₂H₁₃N
MW: 171.24 g/mol
Kokpunkt: 274°C
Flampunkt: 140°C
Storage Temperature: Omgivande
MDL Number: MFCD00003919
CAS nummer: 86-56-6
EINECS: 201-682-4
Merck Index: 14,03252
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b791419ef9f06465233256e7dbf718bd99609f68&vid=b791419ef9f06465233256e7dbf718bd99609f68&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %