3-Hydroxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 3-Hydroxy-naphthalene-2-carboxylic acid, 2-Hydroxy-3-naphthoic acid
H0281-25G H0281-500G
TCIAH0281-25GEA 230 SEK
TCIAH0281-25G TCIAH0281-500G
3-Hydroxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)
3-Hydroxy-2-naphthoic acid
Formel: C₁₁H₈O₃
MW: 188,18 g/mol
Kokpunkt: 368 °C (1013 hPa)
Smältpunkt: 217…220 °C
Flampunkt: 220 °C
MDL Number: MFCD00004103
CAS nummer: 92-70-6
EINECS: 202-180-8
Merck Index: 14,04835
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dae201406d6dec229ea77e336170c4499c70efd2&vid=dae201406d6dec229ea77e336170c4499c70efd2&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 220.0 to 225.0 deg-C
Solubility in Methanol almost transparency