Cyclohexyltrichlorosilane ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Trichloro(cyclohexyl)silane

C0892-25G C0892-5G
TCIAC0892-25GEA 1825 SEK
TCIAC0892-25G TCIAC0892-5G
Cyclohexyltrichlorosilane ≥98.0%
Cyclohexyltrichlorosilane
Formel: C₆H₁₁Cl₃Si
MW: 217,6 g/mol
Kokpunkt: 207°C
Densitet: 1,232 g/cm³ (20 °C)
Flampunkt: 94°C
MDL Number: MFCD00021280
CAS nummer: 98-12-4
EINECS: 202-639-2
UN: 1763
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1726aa17b876b19db5afef2e954cac2436fca968&vid=1726aa17b876b19db5afef2e954cac2436fca968&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR