Phenothiazine ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 10H-Phenothiazine

P0106-25G P0106-500G
TCIAP0106-25GEA 230 SEK
TCIAP0106-25G TCIAP0106-500G
Phenothiazine ≥98.0%
Phenothiazine
Formel: C₁₂H₉NS
MW: 199,28 g/mol
Kokpunkt: 371 °C (1013 hPa)
Smältpunkt: 182…185 °C
Densitet: 1,34 g/cm³ (20 °C)
Flampunkt: 202 °C
MDL Number: MFCD00005015
CAS nummer: 92-84-2
EINECS: 202-196-5
Merck Index: 14,07252
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=571423748a028b43a355533f8ccb01e72eb64ae7&vid=571423748a028b43a355533f8ccb01e72eb64ae7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 183.0 to 188.0 deg-C
Solubility in Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR