1-(4-Nitrophenyl)piperazine ≥98.0%

Leverantör: TCI EUROPE
Varning

N0694-25G N0694-5G
TCIAN0694-25GEA 975 SEK
TCIAN0694-25G TCIAN0694-5G
1-(4-Nitrophenyl)piperazine ≥98.0%
1-(4-Nitrophenyl)piperazine
Formel: C₁₀H₁₃N₃O₂
MW: 207,23 g/mol
Smältpunkt: 136,85 °C
MDL Number: MFCD00005961
CAS nummer: 6269-89-2
EINECS: 228-443-7
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6955a6e9d9ca8f3166efb1eb0a22819c9d0f8a0a&vid=6955a6e9d9ca8f3166efb1eb0a22819c9d0f8a0a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 130.0 to 134.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR