Isoforon ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3,5,5-Trimetyl-2-cyklohexen-1-on

I0151-25ML I0151-500ML
TCIAI0151-25MLEA 215 SEK
TCIAI0151-25ML TCIAI0151-500ML
Isoforon ≥97.0%
Isoforon
Formel: C₉H₁₄O
MW: 138,21 g/mol
Kokpunkt: 215°C
Smältpunkt: –8°C
Densitet: 0,922 g/cm³ (20 °C)
Flampunkt: 84°C
Storage Temperature: Omgivande
MDL Number: MFCD00001584
CAS nummer: 78-59-1
EINECS: 201-126-0
Merck Index: 14,05196
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb9ebd2af76bd750949ebffdcc737e3b6eef2ee2&vid=cb9ebd2af76bd750949ebffdcc737e3b6eef2ee2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR