3-Cyclohexyl-1-propanol ≥99.0%
Leverantör: TCI
Synonyms:
3-Hydroxypropylcyclohexane, 3-Cyclohexylpropan-1-ol
C1166-25ML
C1166-5ML
C1166-100ML
TCIAC1166-25MLEA
975
SEK
TCIAC1166-25ML
TCIAC1166-5ML
TCIAC1166-100ML
3-Cyclohexyl-1-propanol ≥99.0%
3-Cyclohexyl-1-propanol
Formel:
C₉H₁₈O Kokpunkt: 223°C Smältpunkt: –60 °C Densitet: 0,9259 g/cm³ (0 °C) Flampunkt: 101°C |
MDL Number:
MFCD00001528 CAS nummer: 1124-63-6 EINECS: 214-401-5 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...