Tetrabromo-p-benzoquinone ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2,3,5,6-Tetrabromo-1,4-benzoquinone, 2,3,5,6-Tetrabromocyclohexa-2,5-diene-1,4-dione, p-Bromanil, Tetrabromoquinone, 4-Bromanil, Tetrabromo-1,4-benzoquinone

T0617-25G T0617-5G
TCIAT0617-25GEA 1850 SEK
TCIAT0617-25G TCIAT0617-5G
Tetrabromo-p-benzoquinone ≥98.0%
Tetrabromo-p-benzoquinone
Formel: C₆Br₄O₂
MW: 423,68 g/mol
MDL Number: MFCD00013785
CAS nummer: 488-48-2
EINECS: 207-679-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59616a476a74221bad67e76f40f02cba59bf8aeb&vid=59616a476a74221bad67e76f40f02cba59bf8aeb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow powder to crystal
Purity (Cerium redox Method) min. 98.0 %
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR