4-Iodobiphenyl ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Iodo-1,1'-biphenyl
I0785-25G
I0785-5G
TCIAI0785-25GEA
1850
SEK
TCIAI0785-25G
TCIAI0785-5G
4-Iodobiphenyl ≥97.0%
4-Iodobiphenyl
Formel:
C₁₂H₉I MW: 280,11 g/mol Kokpunkt: 320°C Smältpunkt: 112 °C Densitet: 1,584 g/cm³ (20 °C) |
MDL Number:
MFCD00019028 CAS nummer: 1591-31-7 EINECS: 216-469-1 UN: 3077 ADR: 9,III |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 110.0 to 114.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...