Dibenzoylmethane ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 1,3-Diphenyl-1,3-propanedione, 1,3-Diphenylpropane-1,3-dione
D0910-25G D0910-500G D0910-100G
TCIAD0910-25GEA 215 SEK
TCIAD0910-25G TCIAD0910-500G TCIAD0910-100G
Dibenzoylmethane ≥98.0%
Dibenzoylmethane
Formel: (C₆H₅)C(O)CH₂C(O)(C₆H₅)
MW: 224,26 g/mol
Kokpunkt: 357 °C (1013 hPa)
Smältpunkt: 74…77 °C
Densitet: 0,8 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003085
CAS nummer: 120-46-7
EINECS: 204-398-9
Merck Index: 14,03009
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8510dda93a01e8cbb7ebb3a8188c2554887bf54d&vid=8510dda93a01e8cbb7ebb3a8188c2554887bf54d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 80.0 deg-C
Solubility in hot Methanol almost transparency