Oxybenzone ≥99.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Eusolex® 4360, 2-Hydroxy-4-methoxybenzophenone, (2-Hydroxy-4-methoxyphenyl)(phenyl)methanone
H0266-25G H0266-500G
TCIAH0266-25GEA 245 SEK
TCIAH0266-25G TCIAH0266-500G
Oxybenzone ≥99.0%
Oxybenzone
Formel: C₁₄H₁₂O₃
MW: 228,25 g/mol
Kokpunkt: 370 °C (1013 hPa)
Smältpunkt: 65 °C
Densitet: 1,3397 g/cm³ (25 °C)
MDL Number: MFCD00008387
CAS nummer: 131-57-7
EINECS: 205-031-5
UN: 3077
ADR: 9,III
Merck Index: 14,06954
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=deace785d2da5249aa1e6fae6d402108801714cb&vid=deace785d2da5249aa1e6fae6d402108801714cb&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 62.0 to 65.0 deg-C
Solubility in hot Methanol almost transparency