4'-Cyclohexylacetophenone ≥98.0%

Leverantör: TCI
Varning

Synonyms: p-Cyclohexylacetophenone

C1217-25G C1217-5G
TCIAC1217-25GEA 1600 SEK
TCIAC1217-25G TCIAC1217-5G
4'-Cyclohexylacetophenone ≥98.0%
4'-Cyclohexylacetophenone
Formel: C₁₄H₁₈O
Kokpunkt: 128°C
Smältpunkt: 65…67 °C
MDL Number: MFCD00001453
CAS nummer: 18594-05-3
EINECS: 242-432-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=203ea8b1ce8a5f29077481d546122be3c688d724&vid=203ea8b1ce8a5f29077481d546122be3c688d724&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 70.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR