4-Heptylphenol ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
4-n-Heptylphenol
H0615-25G
H0615-5G
TCIAH0615-25GEA
2450
SEK
TCIAH0615-25G
TCIAH0615-5G
4-Heptylphenol ≥98.0%
4-Heptylphenol
Formel:
C₁₃H₂₀O Kokpunkt: 298 °C (1013 hPa) Smältpunkt: 26…28 °C Densitet: 0,939 g/cm³ (20 °C) |
MDL Number:
MFCD00041751 CAS nummer: 1987-50-4 EINECS: 217-862-0 UN: 2430 ADR: 8,III |
|
Specifikation testresultat
Appearance | White or Colorles to Yellow to Orange powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...