α,α-Dimethoxytoluene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Benzaldehyde dimethyl acetal

B1197-25ML B1197-500ML B1197-100ML
TCIAB1197-25MLEA 215 SEK
TCIAB1197-25ML TCIAB1197-500ML TCIAB1197-100ML
α,α-Dimethoxytoluene ≥98.0%
α,α-Dimethoxytoluene
Formel: C₉H₁₂O₂
MW: 152,19 g/mol
Kokpunkt: 196°C
Densitet: 1,01 g/cm³ (20 °C)
Flampunkt: 71°C
MDL Number: MFCD00008491
CAS nummer: 1125-88-8
EINECS: 214-413-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d130e3358305536e7669d4b7650c7727a52fb32&vid=5d130e3358305536e7669d4b7650c7727a52fb32&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR