2-Nitro-4-(trifluoromethyl)anisole
Leverantör: TCI
Fara
Synonyms: 4-Methoxy-3-nitrobenzotrifluoride, 1-Methoxy-2-nitro-4-(trifluoromethyl)benzene
T1042-25G T1042-5G
TCIAT1042-25GEA 885 SEK
TCIAT1042-25G TCIAT1042-5G
2-Nitro-4-(trifluoromethyl)anisole
2-Nitro-4-(trifluoromethyl)anisole
Formel: C₈H₆F₃NO₃
Kokpunkt: 90 - 95 °C (0,5 -rr)
Smältpunkt: 47 °C
MDL Number: MFCD00007099
CAS nummer: 394-25-2
EINECS: 206-891-4
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70652751841d1c2379856d76ec7f4841fdddc604&vid=70652751841d1c2379856d76ec7f4841fdddc604&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 50.0 deg-C
Solubility in Toluene almost transparency