2'-Hydroxy-4'-methoxyacetophenone ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol
H0789-25G H0789-5G
TCIAH0789-25GEA 1050 SEK
TCIAH0789-25G TCIAH0789-5G
2'-Hydroxy-4'-methoxyacetophenone ≥98.0%
2'-Hydroxy-4'-methoxyacetophenone
Formel: C₉H₁₀O₃
MW: 166,18 g/mol
Kokpunkt: 154°C
Smältpunkt: 51 °C
Densitet: 1,1604 g/cm³ (40 °C)
MDL Number: MFCD00008730
CAS nummer: 552-41-0
EINECS: 209-012-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17c1ec5704c528e690870a03103853cef5833892&vid=17c1ec5704c528e690870a03103853cef5833892&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 49.0 to 52.0 deg-C
Solubility in Methanol almost transparency