3-Chloro-4-iodobenzotrifluoride ≥97.0% stabilised
Leverantör: TCI EUROPE
Varning
Stabilization: Stabilised with copper chip
C2336-1G C2336-5G
TCIAC2336-1GEA 245 SEK
TCIAC2336-1G TCIAC2336-5G
3-Chloro-4-iodobenzotrifluoride ≥97.0% stabilised
3-Chloro-4-iodobenzotrifluoride
Formel: C₇H₃ClF₃I
MW: 306,45 g/mol
MDL Number: MFCD00042562
CAS nummer: 141738-80-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69d9628b0ba6ad7bd067efb0cb17fcfd49d9efbc&vid=69d9628b0ba6ad7bd067efb0cb17fcfd49d9efbc&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Purity (GC) min. 97.0 %