3'-Bromo-4'-fluoroacetophenone ≥96.0%

Leverantör: TCI
Varning

B2014-5G B2014-25G
TCIAB2014-5GEA 605 SEK
TCIAB2014-5G TCIAB2014-25G
3'-Bromo-4'-fluoroacetophenone ≥96.0%
3'-Bromo-4'-fluoroacetophenone
Formel: C₈H₆BrFO
Kokpunkt: 240°C
Smältpunkt: 57 °C
MDL Number: MFCD00042466
CAS nummer: 1007-15-4
EINECS: 213-750-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25e573b616de45b58ee85a4588beaad8db763dc0&vid=25e573b616de45b58ee85a4588beaad8db763dc0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 54.0 to 58.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR