ω-Bromo-4-cyanoacetophenone ≥96.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 4-(2-Bromoacetyl)benzonitrile, 4-(Bromoacetyl)benzonitrile, 4-(2-Bromoacetyl)benzenecarbonitrile, 2-Bromo-4'-cyanoacetophenone, Bromomethyl p-cyanophenyl ketone, 2-Bromo-4′-cyanoacetophenone, 4-Cyanophenacyl bromide, α-Bromo-4-cyanoacetophenone

C1348-25G C1348-5G
TCIAC1348-25GEA 1175 SEK
TCIAC1348-25G TCIAC1348-5G
ω-Bromo-4-cyanoacetophenone ≥96.0% (by GC, titration analysis)
ω-Bromo-4-cyanoacetophenone
Formel: C₉H₆BrNO
MW: 224,06 g/mol
Kokpunkt: 314 °C (1013 hPa)
Smältpunkt: 92…94 °C
MDL Number: MFCD00052931
CAS nummer: 20099-89-2
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08195a7c89798206ee29e7c4a57be9485a00d251&vid=08195a7c89798206ee29e7c4a57be9485a00d251&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Purity (Argentmetric Titration) min. 96.0 %
Melting point 92.0 to 94.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR