Dimethyl isophthalate ≥99.0%
Leverantör: TCI
Varning
Synonyms: Isophthalic acid dimethyl ester
I0157-25G I0157-500G
TCIAI0157-25GEA 230 SEK
TCIAI0157-25G TCIAI0157-500G
Dimethyl isophthalate ≥99.0%
Dimethyl isophthalate
Formel: C₁₀H₁₀O₄
MW: 194.19 g/mol
Kokpunkt: 124°C
Storage Temperature: Omgivande
MDL Number: MFCD00008433
CAS nummer: 1459-93-4
EINECS: 215-951-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cd642a8241d6263620a88d10ccb3283b893dffb&vid=5cd642a8241d6263620a88d10ccb3283b893dffb&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 67.0 to 71.0 deg-C
Solubility in Methanol almost transparency