Isophthaloyl dichloride ≥99.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: Isophthaloyl chloride

I0159-500G I0159-25G
TCIAI0159-500GEA 575 SEK
TCIAI0159-500G TCIAI0159-25G
Isophthaloyl dichloride ≥99.0% (by GC, titration analysis)
Isophthaloyl dichloride
Formel: C₈H₄Cl₂O₂
MW: 203,02 g/mol
Kokpunkt: 142°C
Smältpunkt: 42…45 °C
Densitet: 1,388 g/cm³ (17 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00000678
CAS nummer: 99-63-8
EINECS: 202-774-7
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=643a4171e71e4e90ed604c4bbe9f2b913cbbf24d&vid=643a4171e71e4e90ed604c4bbe9f2b913cbbf24d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 99.0 %
Purity (Argentmetric Titration) min. 99.0 %
Melting point 43.0 to 46.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR