2,2,4,4-Tetramethylpentane ≥99.0%
Leverantör: TCI EUROPE
|
Fara
|
T1704-25ML
T1704-5ML
TCIAT1704-25MLEA
2525
SEK
TCIAT1704-25ML
TCIAT1704-5ML
2,2,4,4-Tetramethylpentane ≥99.0%
2,2,4,4-Tetramethylpentane
Formel:
C₉H₂₀ MW: 128,26 g/mol Kokpunkt: 122°C Smältpunkt: –66,5 °C Densitet: 0,7195 g/cm³ (20 °C) Flampunkt: 16°C Storage Temperature: Omgivande |
MDL Number:
MFCD00060873 CAS nummer: 1070-87-7 UN: 1920 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...