Dietylentriamin ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3-Azapentan-1,5-diamin
D0493-25ML
D0493-500ML
TCIAD0493-25MLEA
230
SEK
TCIAD0493-25ML
TCIAD0493-500ML
Dietylentriamin ≥98.0% (by GC, titration analysis)
Dietylentriamin
Formel:
C₄H₁₃N₃ MW: 103,17 g/mol Kokpunkt: 207 °C (1013 hPa) Smältpunkt: –39 °C Densitet: 0,9523 g/cm³ (20 °C) Flampunkt: 97°C Storage Temperature: Omgivande |
MDL Number:
MFCD00008171 CAS nummer: 111-40-0 EINECS: 203-865-4 UN: 2079 ADR: 8,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...