Iron(II) acetate ≥90.0%

Leverantör: TCI EUROPE
Varning

I0765-25G I0765-5G
TCIAI0765-25GEA 1875 SEK
TCIAI0765-25G TCIAI0765-5G
Iron(II) acetate ≥90.0%
Iron(II) acetate
Formel: C₄H₆FeO₄
MW: 173,94 g/mol
Smältpunkt: 190 °C
MDL Number: MFCD00058909
CAS nummer: 3094-87-9
EINECS: 221-441-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65f240c67037c09d18e418d10e1b87d8d5de4a5a&vid=65f240c67037c09d18e418d10e1b87d8d5de4a5a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (Potassium permanganate method) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR