2-Metoxietylacetat ≥98.0%

Leverantör: TCI
Fara

Synonyms: Metylglykolacetat

M0113-25ML M0113-500ML
TCIAM0113-25MLEA 215 SEK
TCIAM0113-25ML TCIAM0113-500ML
2-Metoxietylacetat ≥98.0%
2-Metoxietylacetat
Formel: C₅H₁₀O₃
MW: 118.13 g/mol
Kokpunkt: 143°C
Smältpunkt: -65
Flampunkt: 51°C
MDL Number: MFCD00008719
CAS nummer: 110-49-6
EINECS: 203-772-9
UN: 1189
ADR: 3 III
Merck Index: 14,06039
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a79da10fc207e1d48addb5bcd2faa1e327d3cf8e&vid=a79da10fc207e1d48addb5bcd2faa1e327d3cf8e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR