Triacetin ≥99.0%
Leverantör: TCI
Synonyms: Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate
G0086-25G G0086-500G
TCIAG0086-25GEA 215 SEK
TCIAG0086-25G TCIAG0086-500G
Triacetin ≥99.0%
Triacetin
Formel: C₉H₁₄O₆
MW: 218.21 g/mol
Kokpunkt: 260°C
Smältpunkt: –78°C
Flampunkt: 144°C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00008716
CAS nummer: 102-76-1
EINECS: 203-051-9
Merck Index: 14,09589
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=537a718b750a8c2b0be70aa592df72454a584ea4&vid=537a718b750a8c2b0be70aa592df72454a584ea4&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless clear liquid
Purity (GC) min. 99.0 %
SPECIFIKATIONER
Environmentally Preferable