Trilaurin (glycerol trilaurate) ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Glyceryl tridodecanoate
G0087-25G
G0087-500G
G0087-100G
TCIAG0087-25GEA
415
SEK
TCIAG0087-25G
TCIAG0087-500G
TCIAG0087-100G
Trilaurin (glycerol trilaurate) ≥98.0%
Trilaurin (glycerol trilaurate)
Formel:
C₃₉H₇₄O₆ MW: 639,01 g/mol Kokpunkt: 643 °C (1013 hPa) Smältpunkt: 49 °C Densitet: 0,9713 g/cm³ (20 °C) Storage Temperature: Frys |
MDL Number:
MFCD00026559 CAS nummer: 538-24-9 EINECS: 208-687-0 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 44.0 to 49.0 deg-C |
Solubility in Ethylacetate | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...